Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
twk-8
Entrez Gene ID
177727
Gene Name
protein-coding
Gene Aliases
CELE_ZC410.4
Chromosome Location
Chr.IV: 9082355 - 9087579 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028425.5 NP_001023596.1 12 - 13 2242 116

View Full Product Details