Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rab-28
Entrez Gene ID
189429
Gene Name
Ras-related protein Rab-28
Gene Aliases
CELE_Y11D7A.4
Chromosome Location
Chr.IV: 9242074 - 9244012 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069208.3 NP_501609.1 6 - 7 686 76

View Full Product Details