Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- wrt-3
- Entrez Gene ID
-
177781
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_F38E11.7
- Chromosome Location
-
Chr.IV: 9473633 - 9481011 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001383164.1 |
|
11 - 12 |
1687 |
81 |
- Gene Symbol
- cng-3
- Entrez Gene ID
-
3565463
- Gene Name
- Cyclic nucleotide-binding domain-containing protein
- Gene Aliases
-
CELE_F38E11.12
- Chromosome Location
-
Chr.IV: 9475836 - 9481037 on Build WBcel235
View Full Product Details