Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sru-6
Entrez Gene ID
183142
Gene Name
Serpentine Receptor, class U
Gene Aliases
CELE_C33A12.8
Chromosome Location
Chr.IV: 9490620 - 9491991 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069276.1 NP_501677.1 1 - 2 452 122

View Full Product Details