Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sru-3
Entrez Gene ID
183144
Gene Name
Serpentine Receptor, class U
Gene Aliases
CELE_C33A12.11
Chromosome Location
Chr.IV: 9496606 - 9497888 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069277.1 NP_501678.1 3 - 4 812 103

View Full Product Details