Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dur-1
Entrez Gene ID
177846
Gene Name
protein-coding
Gene Aliases
CELE_F25H8.5
Chromosome Location
Chr.IV: 9921938 - 9928230 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069385.6 NP_501786.2 2 - 3 132 115
NM_069386.8 NP_501787.3 2 - 3 131 115
NM_069387.5 NP_501788.2 2 - 3 132 115

View Full Product Details