Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mspn-1
Entrez Gene ID
177896
Gene Name
protein-coding
Gene Aliases
CELE_K04D7.2
Chromosome Location
Chr.IV: 10177968 - 10180307 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069459.9 NP_501860.1 2 - 3 259 69
NM_069460.2 NP_501861.1 2 - 3 248 69

View Full Product Details