Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
abt-1
Entrez Gene ID
182849
Gene Name
protein-coding
Gene Aliases
CELE_C24F3.5
Chromosome Location
Chr.IV: 10206189 - 10214911 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306925.2 NP_001293854.1 10 - 11 1697 134
NM_001306926.3 NP_001293855.1 7 - 8 1460 134

View Full Product Details