Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- rad-51
- Entrez Gene ID
-
177914
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_Y43C5A.6
- Chromosome Location
-
Chr.IV: 10279462 - 10284252 on Build WBcel235
- Gene Symbol
- thk-1
- Entrez Gene ID
-
177913
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_Y43C5A.5
- Chromosome Location
-
Chr.IV: 10279457 - 10283994 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001383184.1 |
|
2 - 3 |
414 |
74 |
View Full Product Details