Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
unc-43
Entrez Gene ID
177921
Gene Name
protein-coding
Gene Aliases
CELE_K11E8.1
Chromosome Location
Chr.IV: 10324254 - 10350292 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268551.3 NP_001255480.1 13 - 14 1418 89
NM_001268552.3 NP_001255481.1 2 - 3 173 89
NM_001372918.2 NP_001359633.1 13 - 13 1479 89
NM_001380439.2 NP_001366993.1 12 - 13 1386 89
NM_001380440.2 NP_001366994.1 13 - 14 1493 89
NM_001380442.1 NP_001366992.1 10 - 11 1170 89
NM_001383186.1 11 - 12 1246 89
NM_001383187.1 11 - 12 1247 89
GenBank mRNA AF180735.1 - 10 - 11 1148 89
AF233262.1 - 9 - 10 935 89
AF233263.1 - 10 - 11 1088 89
AF233264.1 - 10 - 11 1088 89
AF255956.1 - 9 - 10 935 89

View Full Product Details