Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
unc-43
Entrez Gene ID
177921
Gene Name
protein-coding
Gene Aliases
CELE_K11E8.1
Chromosome Location
Chr.IV: 10324254 - 10350292 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028122.5 NP_001023293.2 2 - 3 172 66
NM_001028127.6 NP_001023298.1 12 - 13 1347 66
NM_001028133.5 NP_001023304.1 2 - 3 657 66
NM_001129316.4 NP_001122788.1 2 - 3 300 66
NM_001136391.4 NP_001129863.1 2 - 3 130 66
NM_001268551.3 NP_001255480.1 14 - 15 1525 66
NM_001268552.3 NP_001255481.1 3 - 4 280 66
NM_001268553.3 NP_001255482.1 2 - 3 376 66
NM_001380439.2 NP_001366993.1 13 - 14 1493 66
NM_001380440.2 NP_001366994.1 14 - 15 1600 66
NM_001383187.1 12 - 13 1354 66
GenBank mRNA AF233262.1 - 10 - 11 1042 66
AF233264.1 - 11 - 12 1195 66
AF233265.1 - 10 - 11 1036 66

View Full Product Details