Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lpd-8
Entrez Gene ID
177928
Gene Name
Nfu_N domain-containing protein
Gene Aliases
CELE_R10H10.1
Chromosome Location
Chr.IV: 10387413 - 10388351 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069516.7 NP_501917.2 2 - 3 123 91

View Full Product Details