Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
him-8
Entrez Gene ID
188247
Gene Name
protein-coding
Gene Aliases
CELE_T07G12.12
Chromosome Location
Chr.IV: 10555646 - 10563975 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383193.1 9 - 10 1914 95

View Full Product Details