Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
efn-2
Entrez Gene ID
177949
Gene Name
Ephrin RBD domain-containing protein
Gene Aliases
CELE_C43F9.8
Chromosome Location
Chr.IV: 10576350 - 10578288 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069554.8 NP_501955.2 2 - 3 325 69

View Full Product Details