Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ncx-3
Entrez Gene ID
177963
Gene Name
Na/Ca eXchangers
Gene Aliases
CELE_ZC168.1
Chromosome Location
Chr.IV: 10716648 - 10721755 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069583.4 NP_501984.2 5 - 6 1253 149

View Full Product Details