Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sdz-13
Entrez Gene ID
184424
Gene Name
SKN-1 Dependent Zygotic transcript
Gene Aliases
CELE_F13E9.6
Chromosome Location
Chr.IV: 10877708 - 10879727 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069617.4 NP_502018.2 6 - 7 681 127

View Full Product Details