Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hrg-4
Entrez Gene ID
185380
Gene Name
protein-coding
Gene Aliases
CELE_F36H1.5
Chromosome Location
Chr.IV: 11030143 - 11030993 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136385.2 NP_001129857.1 2 - 3 344 104

View Full Product Details