Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
str-3
Entrez Gene ID
187459
Gene Name
Seven TM Receptor
Gene Aliases
CELE_M7.13
Chromosome Location
Chr.IV: 11098448 - 11100371 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069670.2 NP_502071.2 2 - 3 299 151

View Full Product Details