Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
spo-11
Entrez Gene ID
191771
Gene Name
protein-coding
Gene Aliases
CELE_T05E11.4
Chromosome Location
Chr.IV: 11123133 - 11124862 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069680.5 NP_502081.1 1 - 2 176 123

View Full Product Details