Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dnj-2
Entrez Gene ID
178043
Gene Name
DnaJ homolog dnj-2
Gene Aliases
CELE_B0035.2
Chromosome Location
Chr.IV: 11312434 - 11313980 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069725.5 NP_502126.1 2 - 3 332 123

View Full Product Details