Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gex-2
Entrez Gene ID
176885
Gene Name
Cytoplasmic FMR1-interacting protein homolog
Gene Aliases
CELE_F56A11.1
Chromosome Location
Chr.IV: 572769 - 583577 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067548.5 NP_499949.2 7 - 8 1021 81
GenBank mRNA AB073209.1 - 7 - 8 1043 81

View Full Product Details