Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srt-23
Entrez Gene ID
190321
Gene Name
Serpentine Receptor, class T
Gene Aliases
CELE_Y55F3C.2
Chromosome Location
Chr.IV: 756721 - 759084 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067578.4 NP_499979.3 1 - 2 172 153

View Full Product Details