Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dcap-1
Entrez Gene ID
176924
Gene Name
mRNA_decap_C domain-containing protein
Gene Aliases
CELE_Y55F3AM.12
Chromosome Location
Chr.IV: 1049836 - 1054794 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067629.6 NP_500030.1 2 - 3 220 104

View Full Product Details