Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srx-14
Entrez Gene ID
3564967
Gene Name
7TM_GPCR_Srx domain-containing protein
Gene Aliases
CELE_C44B12.8
Chromosome Location
Chr.IV: 1111462 - 1113738 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001356906.1 NP_001343637.1 3 - 4 799 122

View Full Product Details