Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-41
Entrez Gene ID
190865
Gene Name
protein-coding
Gene Aliases
CELE_Y104H12A.1
Chromosome Location
Chr.IV: 1386215 - 1400034 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067672.5 NP_500073.3 2 - 3 197 92

View Full Product Details