Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srz-32
Entrez Gene ID
189213
Gene Name
Serpentine Receptor, class Z
Gene Aliases
CELE_W05E7.2
Chromosome Location
Chr.IV: 3436319 - 3437878 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067944.3 NP_500345.2 2 - 3 291 164

View Full Product Details