Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
taf-2
Entrez Gene ID
192051
Gene Name
Transcription initiation factor TFIID subunit 2
Gene Aliases
CELE_Y37E11B.4
Chromosome Location
Chr.IV: 3584298 - 3593613 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001356915.4 NP_001343606.1 1 - 2 249 87

View Full Product Details