Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
map-1
Entrez Gene ID
177128
Gene Name
protein-coding
Gene Aliases
CELE_Y37E11AL.7
Chromosome Location
Chr.IV: 3683537 - 3686865 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067995.4 NP_500396.2 3 - 4 609 147

View Full Product Details