Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
oac-23
Entrez Gene ID
185419
Gene Name
Acyl_transf_3 domain-containing protein
Gene Aliases
CELE_F37C4.3
Chromosome Location
Chr.IV: 3881441 - 3884688 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068030.2 NP_500431.1 9 - 10 1551 125

View Full Product Details