Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
oac-22
Entrez Gene ID
185418
Gene Name
Acyl_transf_3 domain-containing protein
Gene Aliases
CELE_F37C4.2
Chromosome Location
Chr.IV: 3885665 - 3888836 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068031.6 NP_500432.3 8 - 9 1374 97

View Full Product Details