Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
clec-68
Entrez Gene ID
186388
Gene Name
C-type lectin domain-containing protein
Gene Aliases
CELE_F56D6.1
Chromosome Location
Chr.IV: 3926417 - 3929105 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068037.7 NP_500438.5 6 - 7 1149 92

View Full Product Details