Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
clec-78
Entrez Gene ID
185915
Gene Name
C-type LECtin
Gene Aliases
CELE_F47C12.2
Chromosome Location
Chr.IV: 3989102 - 4014235 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068053.5 NP_500454.2 16 - 17 5103 70

View Full Product Details