Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
wrt-5
Entrez Gene ID
177162
Gene Name
protein-coding
Gene Aliases
CELE_W03D2.5
Chromosome Location
Chr.IV: 4050053 - 4052841 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047529.3 NP_001040994.1 2 - 3 282 58

View Full Product Details