Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rnp-3
Entrez Gene ID
177182
Gene Name
protein-coding
Gene Aliases
CELE_K08D10.3
Chromosome Location
Chr.IV: 4174089 - 4176279 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068104.6 NP_500505.1 1 - 2 79 96

View Full Product Details