Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dpy-13
Entrez Gene ID
177187
Gene Name
Cuticle collagen dpy-13
Gene Aliases
CELE_F30B5.1
Chromosome Location
Chr.IV: 4235614 - 4236710 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068118.6 NP_500519.1 1 - 2 99 76

View Full Product Details