Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tni-4
Entrez Gene ID
177295
Gene Name
Troponin I 4
Gene Aliases
CELE_W03F8.1
Chromosome Location
Chr.IV: 5204591 - 5207124 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068340.7 NP_500741.2 3 - 4 427 102

View Full Product Details