Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
elo-6
Entrez Gene ID
177321
Gene Name
Elongation of very long chain fatty acids protein 6
Gene Aliases
CELE_F41H10.8
Chromosome Location
Chr.IV: 5386625 - 5388829 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068396.7 NP_500797.1 3 - 4 840 95

View Full Product Details