Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fem-1
Entrez Gene ID
177335
Gene Name
protein-coding
Gene Aliases
CELE_F35D6.1
Chromosome Location
Chr.IV: 5533140 - 5537992 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068423.6 NP_500824.1 3 - 4 399 64

View Full Product Details