Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
fkh-7
Entrez Gene ID
177340
Gene Name
protein-coding
Gene Aliases
CELE_F26D12.1
Chromosome Location
Chr.IV: 5584669 - 5610378 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027976.6 NP_001023147.1 4 - 5 1350 78
NM_001027977.5 NP_001023148.1 3 - 4 777 78
NM_001306882.4 NP_001293811.1 4 - 5 1335 78
NM_001306883.3 NP_001293812.1 3 - 4 777 78
NM_001306884.4 NP_001293813.1 4 - 5 1335 78
NM_001306885.3 NP_001293814.1 4 - 5 1335 78
NM_001306886.3 NP_001293815.1 3 - 4 777 78
NM_001306887.3 NP_001293816.1 3 - 4 777 78

View Full Product Details