Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-46
Entrez Gene ID
177350
Gene Name
protein-coding
Gene Aliases
CELE_C45E5.6
Chromosome Location
Chr.IV: 5747363 - 5757862 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027894.7 NP_001023065.1 3 - 4 1101 66
NM_001380240.1 NP_001367663.1 2 - 3 511 66
NM_068457.6 NP_500858.2 3 - 4 910 66

View Full Product Details