Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srv-22
Entrez Gene ID
186687
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_H04M03.9
Chromosome Location
Chr.IV: 5878470 - 5879953 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068480.1 NP_500881.1 4 - 5 574 191

View Full Product Details