Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
inx-7
Entrez Gene ID
177364
Gene Name
protein-coding
Gene Aliases
CELE_K02B2.4
Chromosome Location
Chr.IV: 5943981 - 5946638 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306838.3 NP_001293767.1 2 - 3 360 154
NM_068493.8 NP_500894.1 4 - 5 1091 154

View Full Product Details