Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
kcc-2
Entrez Gene ID
177370
Gene Name
protein-coding
Gene Aliases
CELE_H16O14.1
Chromosome Location
Chr.IV: 6040976 - 6074507 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268349.2 NP_001255278.1 7 - 8 1817 116
NM_001268350.4 NP_001255279.2 8 - 9 1836 116
NM_001268351.3 NP_001255280.1 7 - 8 2031 116
NM_001268352.3 NP_001255281.1 7 - 8 1632 116
NM_001380258.1 NP_001368424.1 5 - 6 1464 116
GenBank mRNA FJ593901.1 - 7 - 8 1686 116
FJ593903.1 - 7 - 8 1632 116

View Full Product Details