Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srv-32
Entrez Gene ID
188464
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_T13A10.12
Chromosome Location
Chr.IV: 6279960 - 6282243 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068554.2 NP_500955.1 3 - 4 388 85

View Full Product Details