Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srv-24
Entrez Gene ID
190646
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_Y73B6BL.10
Chromosome Location
Chr.IV: 6288364 - 6290841 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068556.1 NP_500957.1 1 - 2 128 142

View Full Product Details