Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srv-27
Entrez Gene ID
190647
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_Y73B6BL.41
Chromosome Location
Chr.IV: 6296198 - 6298353 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068558.3 NP_500959.2 4 - 5 531 64

View Full Product Details