Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dsl-6
Entrez Gene ID
178063
Gene Name
Delta-like protein
Gene Aliases
CELE_H02I12.4
Chromosome Location
Chr.IV: 11396765 - 11398317 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069747.6 NP_502148.2 4 - 5 527 108

View Full Product Details