Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
wht-2
Entrez Gene ID
178069
Gene Name
ABC transporter domain-containing protein
Gene Aliases
CELE_C10C6.5
Chromosome Location
Chr.IV: 11470436 - 11473422 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069763.7 NP_502164.1 9 - 10 1408 68

View Full Product Details