Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hdl-1
Entrez Gene ID
178129
Gene Name
Probable aromatic-L-amino-acid decarboxylase
Gene Aliases
CELE_ZK829.2
Chromosome Location
Chr.IV: 11946066 - 11949817 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069864.5 NP_502265.2 12 - 13 2351 128

View Full Product Details