Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ddb-1
Entrez Gene ID
178156
Gene Name
DNA damage-binding protein 1
Gene Aliases
CELE_M18.5
Chromosome Location
Chr.IV: 12114810 - 12121192 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069898.8 NP_502299.1 11 - 12 3409 110

View Full Product Details